Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GPRC5C
Entrez Gene ID
697446
Gene Name
G protein-coupled receptor class C group 5 member C
Gene Aliases
-
Chromosome Location
Chr.16: 70771132 - 70791936 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001088824.4 XP_001088824.2 - 1675 121
XM_015120266.2 XP_014975752.1 - 135 121

View Full Product Details