Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RTP4
Entrez Gene ID
708404
Gene Name
receptor transporter protein 4
Gene Aliases
-
Chromosome Location
Chr.2: 10456919 - 10459941 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194237.2 NP_001181166.2 - 269 117
GenBank mRNA JU473043.1 - - 202 117

View Full Product Details