Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MLF1
Entrez Gene ID
704116
Gene Name
myeloid leukemia factor 1
Gene Aliases
-
Chromosome Location
Chr.2: 60832636 - 60869841 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261200.1 NP_001248129.1 - 175 135
GenBank mRNA JU334567.1 - - 163 135
JV047102.1 - - 175 135

View Full Product Details