Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC51
Entrez Gene ID
710311
Gene Name
coiled-coil domain containing 51
Gene Aliases
-
Chromosome Location
Chr.2: 104482808 - 104491211 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015131002.2 XP_014986488.2 - 515 161
XM_015131003.2 XP_014986489.2 - 387 161
XM_015131004.2 XP_014986490.1 - 393 161
XM_028844084.1 XP_028699917.1 - 388 161
XM_028844085.1 XP_028699918.1 - 267 161
GenBank mRNA JU326030.1 - - 200 161
JV639258.1 - - 329 161

View Full Product Details