Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASB7
Entrez Gene ID
693956
Gene Name
ankyrin repeat and SOCS box containing 7
Gene Aliases
EGK_17888
Chromosome Location
Chr.7: 79889545 - 79936215 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266289.2 NP_001253218.1 - 279 111
GenBank mRNA JU333018.1 - - 442 111
JU474371.1 - - 279 111
JV642740.1 - - 489 111

View Full Product Details