Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRAPPC13
Entrez Gene ID
696521
Gene Name
trafficking protein particle complex 13
Gene Aliases
C6H5orf44
Chromosome Location
Chr.6: 64192859 - 64234750 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266356.1 NP_001253285.1 - 465 121
XM_015140114.2 XP_014995600.1 - 621 121
XM_015140115.2 XP_014995601.1 - 927 121
XM_015140116.2 XP_014995602.1 - 621 121
XM_015140117.2 XP_014995603.1 - 348 121
XM_015140118.2 XP_014995604.1 - 389 121
GenBank mRNA JU336462.1 - - 617 121
JU336463.1 - - 617 121
JU336464.1 - - 617 121
JU336465.1 - - 617 121
JU336466.1 - - 617 121
JU472539.1 - - 596 121
JU472540.1 - - 596 121
JV044090.1 - - 465 121
JV044091.1 - - 465 121

View Full Product Details