Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRAPPC13
Entrez Gene ID
696521
Gene Name
trafficking protein particle complex 13
Gene Aliases
C6H5orf44
Chromosome Location
Chr.6: 64192859 - 64234750 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266356.1 NP_001253285.1 - 585 118
XM_015140114.2 XP_014995600.1 - 741 118
XM_015140115.2 XP_014995601.1 - 1047 118
XM_015140116.2 XP_014995602.1 - 741 118
XM_015140117.2 XP_014995603.1 - 468 118
XM_015140118.2 XP_014995604.1 - 509 118
GenBank mRNA JU336462.1 - - 737 118
JU336463.1 - - 737 118
JU336464.1 - - 737 118
JU336465.1 - - 737 118
JU336466.1 - - 737 118
JU472539.1 - - 716 118
JU472540.1 - - 716 118
JV044090.1 - - 585 118
JV044091.1 - - 585 118

View Full Product Details