Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SOST
Entrez Gene ID
713617
Gene Name
sclerostin
Gene Aliases
EGK_08778
Chromosome Location
Chr.16: 54994691 - 54998469 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194608.2 NP_001181537.1 - 243 72

View Full Product Details