Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GJA9
Entrez Gene ID
715648
Gene Name
gap junction protein alpha 9
Gene Aliases
CXNM, EGK_00591, GJA10
Chromosome Location
Chr.1: 185720704 - 185729127 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001113253.4 XP_001113253.2 - 813 137

View Full Product Details