Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LDHAL6B
Entrez Gene ID
702403
Gene Name
lactate dehydrogenase A like 6B
Gene Aliases
-
Chromosome Location
Chr.7: 35627706 - 35630040 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194113.2 NP_001181042.2 - 239 173

View Full Product Details