Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
METTL18
Entrez Gene ID
701890
Gene Name
methyltransferase like 18
Gene Aliases
C1H1orf156, EGK_01952
Chromosome Location
Chr.1: 27122520 - 27124848 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266262.1 NP_001253191.1 - 297 125
GenBank mRNA JU337101.1 - - 336 125
JU477837.1 - - 297 125
JV048694.1 - - 324 125
JV665585.1 - - 351 125

View Full Product Details