Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLRN1
Entrez Gene ID
710133
Gene Name
clarin 1
Gene Aliases
EGK_11997
Chromosome Location
Chr.2: 53101163 - 53144422 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194378.2 NP_001181307.2 - 476 100

View Full Product Details