Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCNV2
Entrez Gene ID
696487
Gene Name
potassium voltage-gated channel modifier subfamily V member 2
Gene Aliases
-
Chromosome Location
Chr.15: 81147932 - 81161155 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001086960.4 XP_001086960.1 - 1859 99

View Full Product Details