Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRSS21
Entrez Gene ID
699296
Gene Name
serine protease 21
Gene Aliases
-
Chromosome Location
Chr.20: 2805252 - 2810128 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015125473.2 XP_014980959.1 - 985 92
GenBank mRNA JU328967.1 - - 325 92
JU479190.1 - - 443 92
JV705693.1 - - 381 92

View Full Product Details