Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKS4B
Entrez Gene ID
697713
Gene Name
ankyrin repeat and sterile alpha motif domain containing 4B
Gene Aliases
-
Chromosome Location
Chr.20: 19948122 - 19976303 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001091739.4 XP_001091739.2 - 268 93

View Full Product Details