Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPATA32
Entrez Gene ID
714131
Gene Name
spermatogenesis associated 32
Gene Aliases
-
Chromosome Location
Chr.16: 56518306 - 56526829 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015119908.2 XP_014975394.1 - 281 64
XM_015119910.2 XP_014975396.1 - 245 64

View Full Product Details