Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM252
Entrez Gene ID
698708
Gene Name
transmembrane protein 252
Gene Aliases
C15H9orf71, C9orf71, EGK_07756
Chromosome Location
Chr.15: 83973094 - 83977343 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193966.1 NP_001180895.1 - 335 157

View Full Product Details