Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCNJ14
Entrez Gene ID
718011
Gene Name
potassium inwardly rectifying channel subfamily J member 14
Gene Aliases
-
Chromosome Location
Chr.19: 48452837 - 48456924 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015124465.2 XP_014979951.1 - 1239 156
GenBank mRNA JU486194.1 - - 633 156

View Full Product Details