Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WFDC13
Entrez Gene ID
717327
Gene Name
WAP four-disulfide core domain 13
Gene Aliases
EGK_02354
Chromosome Location
Chr.10: 80898975 - 80902748 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193446.1 NP_001180375.1 - 89 92

View Full Product Details