Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GIMAP8
Entrez Gene ID
713236
Gene Name
GTPase, IMAP family member 8
Gene Aliases
EGK_14329
Chromosome Location
Chr.3: 176402520 - 176435551 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001098321.4 XP_001098321.2 - 1870 100
XM_015135271.2 XP_014990757.2 - 2297 100
GenBank mRNA JU478245.1 - - 1563 100

View Full Product Details