Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ODF3L1
Entrez Gene ID
706545
Gene Name
outer dense fiber of sperm tails 3 like 1
Gene Aliases
-
Chromosome Location
Chr.7: 52860306 - 52863870 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001104221.4 XP_001104221.1 - 566 84

View Full Product Details