Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FTMT
Entrez Gene ID
699743
Gene Name
ferritin mitochondrial
Gene Aliases
EGK_16780
Chromosome Location
Chr.6: 119408431 - 119409904 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001088733.4 XP_001088733.2 - 761 102

View Full Product Details