Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PHOSPHO1
Entrez Gene ID
698959
Gene Name
phosphoethanolamine/phosphocholine phosphatase 1
Gene Aliases
-
Chromosome Location
Chr.16: 47562390 - 47571053 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001092665.4 XP_001092665.1 - 407 105

View Full Product Details