Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WDR20
Entrez Gene ID
708533
Gene Name
WD repeat domain 20
Gene Aliases
-
Chromosome Location
Chr.7: 164368250 - 164457303 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_028850352.1 XP_028706185.1 - 93 57
XM_028850353.1 XP_028706186.1 - 93 57
XM_028850354.1 XP_028706187.1 - 60 57
XM_028850355.1 XP_028706188.1 - 60 57
XM_028850356.1 XP_028706189.1 - 52 57
XM_028850357.1 XP_028706190.1 - 60 57
XM_028850358.1 XP_028706191.1 - 60 57
XM_028850362.1 XP_028706195.1 - 60 57
XM_028850363.1 XP_028706196.1 - 60 57
XM_028850364.1 XP_028706197.1 - 93 57
XM_028850365.1 XP_028706198.1 - 93 57
XM_028850366.1 XP_028706199.1 - 69 57
XM_028850367.1 XP_028706200.1 - 93 57
XM_028850368.1 XP_028706201.1 - 93 57
GenBank mRNA JU319593.1 - - 37 57
JU319594.1 - - 37 57
JU319595.1 - - 37 57
JV665415.1 - - 41 57

View Full Product Details