Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TMEM37
Entrez Gene ID
695060
Gene Name
transmembrane protein 37
Gene Aliases
-
Chromosome Location
Chr.12: 11691772 - 11703412 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001083482.4 XP_001083482.1 - 621 73
XM_015109929.2 XP_014965415.1 - 1037 73

View Full Product Details