Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SGCA
Entrez Gene ID
704493
Gene Name
sarcoglycan alpha
Gene Aliases
-
Chromosome Location
Chr.16: 46614724 - 46627266 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001092850.4 XP_001092850.1 - 806 154
XM_015119320.2 XP_014974806.1 - 681 154

View Full Product Details