Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MEFV
Entrez Gene ID
701927
Gene Name
MEFV innate immuity regulator, pyrin
Gene Aliases
-
Chromosome Location
Chr.20: 3299141 - 3312946 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001092338.4 XP_001092338.2 - 1894 57

View Full Product Details