Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC25A20
Entrez Gene ID
709029
Gene Name
solute carrier family 25 member 20
Gene Aliases
EGK_11657
Chromosome Location
Chr.2: 104924007 - 104965475 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001265864.1 NP_001252793.1 - 608 151
GenBank mRNA JU321606.1 - - 654 151
JU473369.1 - - 608 151

View Full Product Details