Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IGFBP1
Entrez Gene ID
696994
Gene Name
insulin like growth factor binding protein 1
Gene Aliases
EGK_13713
Chromosome Location
Chr.3: 69477974 - 69483322 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001085935.4 XP_001085935.2 - 981 111

View Full Product Details