Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SERPINA5
Entrez Gene ID
702078
Gene Name
serpin family A member 5
Gene Aliases
-
Chromosome Location
Chr.7: 156750129 - 156762138 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044733.3 NP_001038198.3 - 1098 101
GenBank mRNA U22417.1 - - 886 101

View Full Product Details