Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ACHE
Entrez Gene ID
713370
Gene Name
acetylcholinesterase
Gene Aliases
-
Chromosome Location
Chr.3: 42972754 - 42981090 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128088.2 NP_001121560.2 - 1839 143
XM_028845022.1 XP_028700855.1 - 2668 143
XM_028845027.1 XP_028700860.1 - 2191 143
XM_028845028.1 XP_028700861.1 - 2190 143
XM_028845029.1 XP_028700862.1 - 2993 143
XM_028845030.1 XP_028700863.1 - 2134 143
GenBank mRNA JU327112.1 - - 1632 143
JV582285.1 - - 1789 143
JV617130.1 - - 1391 143
JV715348.1 - - 1839 143

View Full Product Details