Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCKAR
Entrez Gene ID
694742
Gene Name
cholecystokinin A receptor
Gene Aliases
-
Chromosome Location
Chr.5: 25444936 - 25454427 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001084186.4 XP_001084186.1 - 788 94

View Full Product Details