Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CD3D
Entrez Gene ID
699582
Gene Name
CD3d molecule
Gene Aliases
-
Chromosome Location
Chr.14: 111394605 - 111399738 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015115817.2 XP_014971303.1 - 2068 96
XM_015115818.2 XP_014971304.1 - 2053 96
GenBank mRNA JV726677.1 - - 263 96

View Full Product Details