Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FOLR2
Entrez Gene ID
718405
Gene Name
folate receptor beta
Gene Aliases
EGK_06646
Chromosome Location
Chr.14: 65188109 - 65193291 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260645.1 NP_001247574.1 - 620 140
XM_015115128.2 XP_014970614.1 - 656 140
XM_028832851.1 XP_028688684.1 - 477 140
GenBank mRNA JU472594.1 - - 620 140

View Full Product Details