Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GABRA1
Entrez Gene ID
574302
Gene Name
gamma-aminobutyric acid type A receptor subunit alpha1
Gene Aliases
EGK_17100
Chromosome Location
Chr.6: 159259664 - 159315700 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260932.1 NP_001247861.1 - 273 114
XM_015141354.2 XP_014996840.1 - 654 114
XM_028849019.1 XP_028704852.1 - 301 114
XM_028849020.1 XP_028704853.1 - 551 114
XM_028849021.1 XP_028704854.1 - 309 114
GenBank mRNA JU320577.1 - - 651 114
JU320578.1 - - 273 114
JV703245.1 - - 687 114

View Full Product Details