Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GRM2
Entrez Gene ID
700214
Gene Name
glutamate metabotropic receptor 2
Gene Aliases
EGK_11587
Chromosome Location
Chr.2: 107787490 - 107799612 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001265680.1 NP_001252609.1 - 611 110
XM_015130559.2 XP_014986045.2 - 2241 110
XM_015130560.2 XP_014986046.2 - 2241 110
XM_028843161.1 XP_028698994.1 - 1217 110
GenBank mRNA JV566976.1 - - 689 110
JV667261.1 - - 611 110

View Full Product Details