Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
REEP3
Entrez Gene ID
697494
Gene Name
receptor accessory protein 3
Gene Aliases
-
Chromosome Location
Chr.9: 74345476 - 74448401 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261663.1 NP_001248592.1 - 409 122
GenBank mRNA JU333391.1 - - 464 122
JU472949.1 - - 393 122
JV046449.1 - - 409 122

View Full Product Details