Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PIM3
Entrez Gene ID
720591
Gene Name
Pim-3 proto-oncogene, serine/threonine kinase
Gene Aliases
-
Chromosome Location
Chr.10: 1506224 - 1509779 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015150605.2 XP_015006091.1 - 646 118
GenBank mRNA JU322603.1 - - 244 118
JU471174.1 - - 378 118

View Full Product Details