Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EXOSC3
Entrez Gene ID
716347
Gene Name
exosome component 3
Gene Aliases
-
Chromosome Location
Chr.15: 46328427 - 46348893 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257940.2 NP_001244869.1 - 334 67
XM_015117402.2 XP_014972888.1 - 729 67
XM_015117403.2 XP_014972889.1 - 730 67
XM_015117404.2 XP_014972890.1 - 759 67
GenBank mRNA JU322413.1 - - 334 67
JV685678.1 - - 354 67

View Full Product Details