Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CENPM
Entrez Gene ID
709006
Gene Name
centromere protein M
Gene Aliases
EGK_03101
Chromosome Location
Chr.10: 9557825 - 9567515 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194334.2 NP_001181263.1 - 114 101
XM_028827317.1 XP_028683150.1 - 142 101

View Full Product Details