Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RABL2B
Entrez Gene ID
716615
Gene Name
RAB, member of RAS oncogene family like 2B
Gene Aliases
-
Chromosome Location
Chr.10: 703286 - 717663 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260670.2 NP_001247599.1 - 260 74
XM_015150634.2 XP_015006120.2 - 623 74
XM_015150635.2 XP_015006121.2 - 733 74
XM_015150638.2 XP_015006124.2 - 377 74
XM_028827398.1 XP_028683231.1 - 881 74
XM_028827399.1 XP_028683232.1 - 877 74
XM_028827400.1 XP_028683233.1 - 595 74
XM_028827401.1 XP_028683234.1 - 486 74
XM_028827402.1 XP_028683235.1 - 771 74
XM_028827403.1 XP_028683236.1 - 617 74
XM_028827404.1 XP_028683237.1 - 158 74
XM_028827406.1 XP_028683239.1 - 626 74
XM_028827407.1 XP_028683240.1 - 625 74
XM_028827408.1 XP_028683241.1 - 378 74
XM_028827409.1 XP_028683242.1 - 629 74
GenBank mRNA JU323199.1 - - 343 74
JU471428.1 - - 268 74
JV046060.1 - - 260 74
JV567305.1 - - 406 74
JV704339.1 - - 487 74

Gene Symbol
LOC114677631
Entrez Gene ID
114677631
Gene Name
rab-like protein 2B
Gene Aliases
-
Chromosome Location
Chr.4: 136662548 - 136663764 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_028847684.1 XP_028703517.1 - 296 74

View Full Product Details