Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OR4K1
Entrez Gene ID
698752
Gene Name
olfactory receptor, family 4, subfamily K, member 1
Gene Aliases
-
Chromosome Location
Chr.7: 81760121 - 81789254 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193794.2 NP_001180723.2 - 894 116
XM_015143083.2 XP_014998569.2 - 2229 116

View Full Product Details