Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RGS10
Entrez Gene ID
703125
Gene Name
regulator of G protein signaling 10
Gene Aliases
-
Chromosome Location
Chr.9: 119981531 - 120024459 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261654.1 NP_001248583.1 - 416 82
XM_015148166.2 XP_015003652.1 - 480 82
GenBank mRNA JU335788.1 - - 464 82
JU335789.1 - - 416 82
JU490991.1 - - 449 82

View Full Product Details