Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLEC6A
Entrez Gene ID
716162
Gene Name
C-type lectin domain containing 6A
Gene Aliases
-
Chromosome Location
Chr.11: 11339495 - 11361755 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190740.1 NP_001177669.1 - 368 74

View Full Product Details