Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IRAK1BP1
Entrez Gene ID
716772
Gene Name
interleukin 1 receptor associated kinase 1 binding protein 1
Gene Aliases
EGK_15099
Chromosome Location
Chr.4: 94155250 - 94190531 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194610.2 NP_001181539.1 - 339 125
GenBank mRNA JU480493.1 - - 253 125

View Full Product Details