Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLVS2
Entrez Gene ID
713626
Gene Name
clavesin 2
Gene Aliases
EGK_15384, clavesin-2
Chromosome Location
Chr.4: 26518641 - 26594053 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194395.2 NP_001181324.1 - 869 118
XM_015137306.2 XP_014992792.1 - 1923 118
GenBank mRNA JU320780.1 - - 743 118
JV452777.1 - - 743 118
JV489895.1 - - 749 118

View Full Product Details