Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ENHO
Entrez Gene ID
701072
Gene Name
energy homeostasis associated
Gene Aliases
EGK_07602
Chromosome Location
Chr.15: 49672689 - 49675202 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261591.1 NP_001248520.1 - 91 54
XM_028834261.1 XP_028690094.1 - 792 54
GenBank mRNA JU320763.1 - - 173 54
JV043715.1 - - 91 54
JV566639.1 - - 265 54

View Full Product Details