Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OTUD6A
Entrez Gene ID
694498
Gene Name
OTU deubiquitinase 6A
Gene Aliases
DUBA-2, EGK_20605
Chromosome Location
Chr.X: 67109949 - 67110815 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193698.2 NP_001180627.1 - 625 120

View Full Product Details