Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C7H14orf28
Entrez Gene ID
701956
Gene Name
chromosome 7 C14orf28 homolog
Gene Aliases
C14orf28, EGK_18148
Chromosome Location
Chr.7: 107184024 - 107194375 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190833.1 NP_001177762.1 - 740 150
XM_015143609.2 XP_014999095.1 - 1041 150
XM_015143610.2 XP_014999096.1 - 926 150
GenBank mRNA JU332874.1 - - 759 150
JV641918.1 - - 774 150

View Full Product Details