Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MSS51
Entrez Gene ID
707066
Gene Name
MSS51 mitochondrial translational activator
Gene Aliases
ZMYND17
Chromosome Location
Chr.9: 64281761 - 64293825 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015147379.2 XP_015002865.2 - 2593 100
XM_015147380.2 XP_015002866.2 - 2749 100
XM_015147381.2 XP_015002867.2 - 2669 100
GenBank mRNA JU479921.1 - - 427 100

View Full Product Details